APO517333953 Apollo Scientific Ltd

4-Methyl-2-nitrophenyl isocyanate

  • MDL number MFCD00014705
  • Cas Number : 57910-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-58
Purity 97
Flash Point 113
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methyl-2-nitrophenyl-isocyanate

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-methyl-2-nitrophenyl-isocyanate
pictograms GHS07;GHS08
hstatements H302+H312+H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart