APO517335703 Apollo Scientific Ltd

2-(Aminomethyl)-1,3-oxazole-4-carboxylic acid, 2-BOC protected

  • MDL number MFCD08236858
  • Cas Number : 182120-90-7
  • Molecular Weight : 242.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-{[(tert-butoxycarbonyl)amino]methyl}-1,3-oxazole-4-carboxylic acid
Inchi Code 1S/C10H14N2O5/c1-10(2,3)17-9(15)11-4-7-12-6(5-16-7)8(13)14/h5H,4H2,1-3H3,(H,11,15)(H,13,14)
Inchi Key AKUIDVVQFDMLAW-UHFFFAOYSA-N

Description

Synonym 2-{[(tert-Butoxycarbonyl)amino]methyl}-1,3-oxazole-4-carboxylic acid
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-aminomethyl-13-oxazole-4-carboxylic-acid-2-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-aminomethyl-13-oxazole-4-carboxylic-acid-2-boc-protected
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart