APO517335707 Apollo Scientific Ltd

3,6-Diiodo-carbazole

  • MDL number MFCD00450017
  • Cas Number : 57103-02-3
  • Molecular Weight : 419

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 207
Iupac Name 3,6-diiodo-9H-carbazole
Inchi Code 1S/C12H7I2N/c13-7-1-3-11-9(5-7)10-6-8(14)2-4-12(10)15-11/h1-6,15H
Inchi Key PECAOKZHORDWAI-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 207
Product Link https://store.apolloscientific.co.uk/product/36-diiodo-carbazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/36-diiodo-carbazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart