APO517335709 Apollo Scientific Ltd

1-Methyl-1H-indole-4-carbonitrile

  • MDL number MFCD08690265
  • Cas Number : 628711-58-0
  • Molecular Weight : 156.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 41-43.5
Iupac Name 1-methyl-1H-indole-4-carbonitrile
Inchi Code 1S/C10H8N2/c1-12-6-5-9-8(7-11)3-2-4-10(9)12/h2-6H,1H3
Inchi Key DYOVHBKIEBCIIE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 41-43.5
Product Link https://store.apolloscientific.co.uk/product/1-methyl-1h-indole-4-carbonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-1h-indole-4-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart