APO630168846 Apollo Scientific Ltd

4-Bromo-2-(trifluoromethoxy)phenylacetylene

  • MDL number MFCD22570386
  • Cas Number : 1824630-55-8
  • Molecular Weight : 265.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-1-ethynyl-2-(trifluoromethoxy)benzene
Inchi Code 1S/C9H4BrF3O/c1-2-6-3-4-7(10)5-8(6)14-9(11,12)13/h1,3-5H
Inchi Key CXEQOKLPHZGESS-UHFFFAOYSA-N

Description

Synonym 4-Bromo-1-ethynyl-2-(trifluoromethoxy)benzene
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethoxyphenylacetylene

Safety Information

signalword Warning
pcodes P210;P280;P303+P361+P353
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethoxyphenylacetylene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart