APO631903447 Apollo Scientific Ltd

3-Iodo-2-nitroanisole

  • MDL number MFCD18397933
  • Cas Number : 725266-66-0
  • Molecular Weight : 279.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-83
Synonym 1-Iodo-3-methoxy-2-nitrobenzene, 3-Iodo-2-nitrophenyl methyl ether
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-iodo-2-nitroanisole

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-iodo-2-nitroanisole
pictograms GHS07;GHS08
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart