APO631903451 Apollo Scientific Ltd

4-Bromo-6-methylpyridazin-3(2H)-one

  • MDL number MFCD09701342
  • Cas Number : 954240-46-1
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-6-methyl-3(2H)-pyridazinone
Inchi Code 1S/C5H5BrN2O/c1-3-2-4(6)5(9)8-7-3/h2,7H,1H2,(H,8,9)
Inchi Key AVNYZCADPOWKPE-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-6-methylpyridazin-32h-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-6-methylpyridazin-32h-one
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart