APO631903453 Apollo Scientific Ltd

2-Bromo-6-iodophenol

  • MDL number MFCD16999870
  • Cas Number : 2040-86-0
  • Molecular Weight : 298.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 60-63
Iupac Name 2-bromo-6-iodophenol
Inchi Code 1S/C6H4BrIO/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
Inchi Key ZZBRPSNXUDZTRX-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 60-63
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-iodophenol

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-iodophenol
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart