APO631903457 Apollo Scientific Ltd

2-Bromo-4-(trifluoromethyl)benzaldehyde

  • MDL number MFCD09835101
  • Cas Number : 85118-24-7
  • Molecular Weight : 253.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 92
Iupac Name 2-bromo-4-(trifluoromethyl)benzaldehyde
Inchi Code 1S/C8H4BrF3O/c9-7-3-6(8(10,11)12)2-1-5(7)4-13/h1-4H
Inchi Key CUKSTNNYAHZPRM-UHFFFAOYSA-N

Description

Melting Point 92
Synonym 3-Bromo-4-formylbenzotrifluoride
Flash Point 229
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-trifluoromethylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-trifluoromethylbenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart