APO631903467 Apollo Scientific Ltd

Methyl 2,4-difluoro-5-iodobenzoate

  • MDL number MFCD24107099
  • Cas Number : 1113050-16-0
  • Molecular Weight : 298.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2,4-difluoro-5-iodobenzoate
Inchi Code 1S/C8H5F2IO2/c1-13-8(12)4-2-7(11)6(10)3-5(4)9/h2-3H,1H3
Inchi Key RKMKPEBEUQDDMM-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-24-difluoro-5-iodobenzoate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/methyl-24-difluoro-5-iodobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart