APO633226112 Apollo Scientific Ltd

(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid

  • MDL number MFCD06245370
  • Cas Number : 195202-08-5
  • Molecular Weight : 254.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-2-(3,4,5-trimethoxyphenyl)butanoic acid
Inchi Code 1S/C13H18O5/c1-5-9(13(14)15)8-6-10(16-2)12(18-4)11(7-8)17-3/h6-7,9H,5H2,1-4H3,(H,14,15)/t9-/m0/s1
Inchi Key WBULUDGUWZFLMO-VIFPVBQESA-N

Description

Synonym (2S)-2-(3,4,5-Trimethoxyphenyl)butyric acid
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2s-2-345-trimethoxyphenylbutanoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2s-2-345-trimethoxyphenylbutanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart