APO715369867 Apollo Scientific Ltd

Methyl 2-Methyl-3-(methylsulfonyl)benzoate

  • MDL number MFCD18071000
  • Cas Number : 1256633-15-4
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-methyl-3-(methylsulfonyl)benzoate
Inchi Code 1S/C10H12O4S/c1-7-8(10(11)14-2)5-4-6-9(7)15(3,12)13/h4-6H,1-3H3
Inchi Key XHFQPCDSXMVFMZ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-methyl-3-methylsulfonylbenzoate

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/methyl-2-methyl-3-methylsulfonylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart