APO715370533 Apollo Scientific Ltd

3-(Hydroxymethyl)-4-methylphenol

  • MDL number MFCD18397253
  • Cas Number : 1261454-85-6
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(hydroxymethyl)-4-methylphenol
Inchi Code 1S/C8H10O2/c1-6-2-3-8(10)4-7(6)5-9/h2-4,9-10H,5H2,1H3
Inchi Key XBPRAIIIFOIDQE-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-hydroxymethyl-4-methylphenol

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxymethyl-4-methylphenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart