APO715371222 Apollo Scientific Ltd

2-Bromo-6-(2-hydroxyethyl)pyridine

  • MDL number MFCD10000073
  • Cas Number : 955370-07-7
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-2-hydroxyethylpyridine

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-2-hydroxyethylpyridine
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart