APO715371415 Apollo Scientific Ltd

1-Methylimidazole-5-boronic acid Pinacol Ester

  • MDL number MFCD11846349
  • Cas Number : 942070-72-6
  • Molecular Weight : 208.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole
Inchi Code 1S/C10H17BN2O2/c1-9(2)10(3,4)15-11(14-9)8-6-12-7-13(8)5/h6-7H,1-5H3
Inchi Key SNKPPKAANOYIRX-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-methylimidazole-5-boronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-methylimidazole-5-boronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart