APO715371764 Apollo Scientific Ltd

(2,4-Difluorophenyl)trimethylsilane

  • MDL number MFCD22682801
  • Cas Number : 148854-10-8
  • Molecular Weight : 186.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-difluorophenyl)trimethylsilane
density 1.049
Inchi Code 1S/C9H12F2Si/c1-12(2,3)9-5-4-7(10)6-8(9)11/h4-6H,1-3H3
Inchi Key CVMLAPBCXNIHBT-UHFFFAOYSA-N

Description

Density 1.049
Flash Point 166-167/747mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluorophenyltrimethylsilane

Safety Information

signalword Warning
pcodes P210
MSDS https://store.apolloscientific.co.uk/product/24-difluorophenyltrimethylsilane
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart