APO715372455 Apollo Scientific Ltd

3-bromo-4-chloro-5-fluorobenzoic acid

  • Molecular Weight : 253.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-4-chloro-5-fluorobenzoic acid
Inchi Code 1S/C7H3BrClFO2/c8-4-1-3(7(11)12)2-5(10)6(4)9/h1-2H,(H,11,12)
Inchi Key CLXLNVHYXWMAAZ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-chloro-5-fluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart