APO715372459 Apollo Scientific Ltd

Methyl 3-bromo-4-chloro-5-fluorobenzoate

  • MDL number MFCD11846086
  • Cas Number : 1160574-62-8
  • Molecular Weight : 267.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-bromo-4-chloro-5-fluorobenzoate
Inchi Code 1S/C8H5BrClFO2/c1-13-8(12)4-2-5(9)7(10)6(11)3-4/h2-3H,1H3
Inchi Key XRJVAGJSNGAQKU-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-bromo-4-chloro-5-fluorobenzoate

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/methyl-3-bromo-4-chloro-5-fluorobenzoate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart