APO715372510 Apollo Scientific Ltd

Methyl 3-iodo-5-(trifluoromethyl)benzoate

  • MDL number MFCD11976042
  • Cas Number : 161622-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid-Oil
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=105341

Safety Information

signalword Warning
pcodes P260;P280;P301;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=105341
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart