APO715372515 Apollo Scientific Ltd

5-Fluoro-2-(hydroxymethyl)benzeneboronic acid

  • MDL number MFCD11111910
  • Cas Number : 1246633-53-3
  • Molecular Weight : 169.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-2-(hydroxymethyl)phenylboronic acid
Inchi Code 1S/C7H8BFO3/c9-6-2-1-5(4-10)7(3-6)8(11)12/h1-3,10-12H,4H2
Inchi Key OBSMVIAXTKAJFM-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-hydroxymethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-hydroxymethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart