APO725185996 Apollo Scientific Ltd

2-Amino-5-bromothiophenol

  • MDL number MFCD00839467
  • Cas Number : 23451-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-115
Synonym 2-Amino-5-bromobenzenethiol, 2-Amino-5-bromophenyl mercaptan
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=73045

Safety Information

signalword Danger
pcodes P260;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=73045
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart