APO725187694 Apollo Scientific Ltd

5-Amino-4-chloro-6-methoxy-2-methylpyrimidine

  • MDL number MFCD01837887
  • Cas Number : 88474-31-1
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-6-methoxy-2-methyl-5-pyrimidinamine
Inchi Code 1S/C6H8ClN3O/c1-3-9-5(7)4(8)6(10-3)11-2/h8H2,1-2H3
Inchi Key XOPAFBPYRUSQNI-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-4-chloro-6-methoxy-2-methylpyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-4-chloro-6-methoxy-2-methylpyrimidine
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart