APO725187695 Apollo Scientific Ltd

Methyl 4-bromo-2-cyanobenzoate

  • MDL number MFCD18398741
  • Cas Number : 1223434-15-8
  • Molecular Weight : 240.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-bromo-2-cyanobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-bromo-2-cyanobenzoate
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart