APO725187701 Apollo Scientific Ltd

8-Bromo-5-nitroisoquinoline

  • MDL number MFCD08282783
  • Cas Number : 252861-41-9
  • Molecular Weight : 253.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-bromo-5-nitroisoquinoline

Safety Information

signalword Warning
pcodes P280;P271
MSDS https://store.apolloscientific.co.uk/product/8-bromo-5-nitroisoquinoline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart