APO725196390 Apollo Scientific Ltd

3-(Difluoromethyl)benzeneboronic acid

  • MDL number MFCD09864670
  • Cas Number : 854690-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38-40
Flash Point 113
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 312/ 760mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40853

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40853
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart