APO725203149 Apollo Scientific Ltd

4-Hydroxy-3-methoxybenzotrifluoride

  • MDL number MFCD13185693
  • Cas Number : 1027888-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methoxy-4-(trifluoromethyl)phenol
Storage temp. Store at 2-8°C
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-3-methoxybenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-3-methoxybenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart