APO725205534 Apollo Scientific Ltd

3-Bromo-2-(trifluoromethyl)benzaldehyde

  • MDL number MFCD22566297
  • Cas Number : 1289057-68-6
  • Molecular Weight : 253.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-trifluoromethylbenzaldehyde

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-trifluoromethylbenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart