APO917953769 Apollo Scientific Ltd

6-Hydroxy-4-methylcoumarin

  • MDL number MFCD00016967
  • Cas Number : 2373-31-1
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 240-241
Iupac Name 6-hydroxy-4-methyl-2H-chromen-2-one
Inchi Code 1S/C10H8O3/c1-6-4-10(12)13-9-3-2-7(11)5-8(6)9/h2-5,11H,1H3
Inchi Key IRUHWRSITUYICV-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 240-241
Product Link https://store.apolloscientific.co.uk/product/6-hydroxy-4-methylcoumarin

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-hydroxy-4-methylcoumarin
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart