APO917953772 Apollo Scientific Ltd

3-(1,3-Benzodioxol-5-yl)-1-phenylprop-2-en-1-one

  • MDL number MFCD00022990
  • Cas Number : 644-34-8
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123
Purity 99
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-13-benzodioxol-5-yl-1-phenylprop-2-en-1-one

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-13-benzodioxol-5-yl-1-phenylprop-2-en-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart