APO917953779 Apollo Scientific Ltd

6-Hydroxychromene-3-carboxaldehyde

  • MDL number MFCD03548123
  • Cas Number : 134822-76-7
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-169
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-hydroxychromene-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340;P305+P351+P338;P312;P321
MSDS https://store.apolloscientific.co.uk/product/6-hydroxychromene-3-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart