APO917953785 Apollo Scientific Ltd

5-Bromo-2-(trifluoromethyl)-1,3-benzoxazole

  • MDL number MFCD03095019
  • Cas Number : 375369-08-7
  • Molecular Weight : 266.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethyl-13-benzoxazole

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethyl-13-benzoxazole
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart