APO945298482 Apollo Scientific Ltd

N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide

  • MDL number MFCD01567008
  • Cas Number : 108825-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/n-methyl-n-3-3-methyl124triazolo43-bpyridazin-6-ylphenylacetamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-methyl-n-3-3-methyl124triazolo43-bpyridazin-6-ylphenylacetamide
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart