APO945298536 Apollo Scientific Ltd

1,3-Bis(hydroxymethyl)-5,5-dimethylhydantoin

  • MDL number MFCD00467199
  • Cas Number : 6440-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-88
Synonym 1,3-Bis(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione, 1,3-Bis(hydroxymethyl)-4,4-dimethyl-2,5-dioxoimidazolidine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42855

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42855
pictograms GHS07
hstatements H315;H317;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart