APO945298549 Apollo Scientific Ltd

4-Bromo-2,6-dinitrotoluene

  • MDL number MFCD07357280
  • Cas Number : 95192-64-6
  • Molecular Weight : 261.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 86-89
Iupac Name 5-bromo-2-methyl-1,3-dinitrobenzene
Inchi Code 1S/C7H5BrN2O4/c1-4-6(9(11)12)2-5(8)3-7(4)10(13)14/h2-3H,1H3
Inchi Key UOGCLPDKGPPDHM-UHFFFAOYSA-N

Description

Synonym 5-Bromo-1,3-dinitro-2-methylbenzene
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 86-89
Product Link https://store.apolloscientific.co.uk/product/4-bromo-26-dinitrotoluene

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-26-dinitrotoluene
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart