APO945298555 Apollo Scientific Ltd

2-Nitrophenylacetylene

  • MDL number MFCD01113917
  • Cas Number : 16433-96-8
  • Molecular Weight : 147.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 82-85
Iupac Name 1-ethynyl-2-nitrobenzene
Inchi Code 1S/C8H5NO2/c1-2-7-5-3-4-6-8(7)9(10)11/h1,3-6H
Inchi Key FWAGYANFBMIHFQ-UHFFFAOYSA-N

Description

Synonym 1-Ethynyl-2-nitrobenzene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 82-85
Product Link https://store.apolloscientific.co.uk/product/2-nitrophenylacetylene

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/2-nitrophenylacetylene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart