APO945298557 Apollo Scientific Ltd

5-Chloro-2-ethoxynicotinic acid

  • MDL number MFCD07403147
  • Cas Number : 68359-07-9
  • Molecular Weight : 201.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 115.5-117.5
Iupac Name 5-chloro-2-ethoxynicotinic acid
Inchi Code 1S/C8H8ClNO3/c1-2-13-7-6(8(11)12)3-5(9)4-10-7/h3-4H,2H2,1H3,(H,11,12)
Inchi Key DTIBPOFHVJBUSH-UHFFFAOYSA-N

Description

Synonym 5-Chloro-2-ethoxypyridine-3-carboxylic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 115.5-117.5
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-ethoxynicotinic-acid

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-ethoxynicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart