APO945298704 Apollo Scientific Ltd

5-Amino-2-methyl-2H-1,4-benzoxazin-3(4H)-one

  • MDL number MFCD11915246
  • Cas Number : 1176714-94-5
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-184
Synonym 5-Amino-3,4-dihydro-2-methyl-3-oxo-2H-1,4-benzoxazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-amino-2-methyl-2h-14-benzoxazin-34h-one

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-2-methyl-2h-14-benzoxazin-34h-one
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart