APO945298708 Apollo Scientific Ltd

6-Chloropyridazine-3-carboxaldehyde

  • MDL number MFCD10698041
  • Cas Number : 303085-53-2
  • Molecular Weight : 142.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-3-pyridazinecarbaldehyde
Inchi Code 1S/C5H3ClN2O/c6-5-2-1-4(3-9)7-8-5/h1-3H
Inchi Key YDHRPQYMQCOQQT-UHFFFAOYSA-N

Description

Synonym 6-Chloro-3-formylpyridazine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloropyridazine-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloropyridazine-3-carboxaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart