APO945298730 Apollo Scientific Ltd

4-Bromophthalic anhydride

  • MDL number MFCD00191323
  • Cas Number : 86-90-8
  • Molecular Weight : 227.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 107
Iupac Name 5-bromo-2-benzofuran-1,3-dione
Inchi Code 1S/C8H3BrO3/c9-4-1-2-5-6(3-4)8(11)12-7(5)10/h1-3H
Inchi Key BCKVHOUUJMYIAN-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-benzofuran-1,3-dione, 5-Bromoisobenzofuran-1,3-dione
Purity 98%
Flash Point 301
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 107
Product Link https://store.apolloscientific.co.uk/product/4-bromophthalic-anhydride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromophthalic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart