APO945298767 Apollo Scientific Ltd

4-(Bromomethyl)benzyl alcohol

  • MDL number MFCD08275218
  • Cas Number : 71831-21-5
  • Molecular Weight : 201.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 76-77
Iupac Name [4-(bromomethyl)phenyl]methanol
Inchi Code 1S/C8H9BrO/c9-5-7-1-3-8(6-10)4-2-7/h1-4,10H,5-6H2
Inchi Key RDHSYXFAOVTAEH-UHFFFAOYSA-N

Description

Synonym [4-(Bromomethyl)phenyl]methanol
Purity 97%
Storage temp. Store at -20
Physical Form Solid-Powder
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 76-77
Product Link https://store.apolloscientific.co.uk/product/4-bromomethylbenzyl-alcohol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromomethylbenzyl-alcohol
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart