APO945298792 Apollo Scientific Ltd

Methyl 5-phenylnicotinate

  • MDL number MFCD06802742
  • Cas Number : 10177-13-6
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 356/760mm
Product Link https://store.apolloscientific.co.uk/product/methyl-5-phenylnicotinate

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-phenylnicotinate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart