APO945298869 Apollo Scientific Ltd

3,4-Dimethyloctane

  • MDL number MFCD30534471
  • Cas Number : 15869-92-8
  • Molecular Weight : 142.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/34-dimethyloctane

Safety Information

signalword Warning
pcodes P210;P260
MSDS https://store.apolloscientific.co.uk/product/34-dimethyloctane
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart