APO945298870 Apollo Scientific Ltd

2,4-Dimethyloctane

  • MDL number MFCD30534470
  • Cas Number : 4032-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 152-153
Product Link https://store.apolloscientific.co.uk/product/24-dimethyloctane

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/24-dimethyloctane
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart