APO945298897 Apollo Scientific Ltd

6-Bromo-1-methylindazole-3-carboxylic acid

  • MDL number MFCD15071436
  • Cas Number : 1021859-29-9
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-1-methyl-1H-indazole-3-carboxylic acid
Inchi Code 1S/C9H7BrN2O2/c1-12-7-4-5(10)2-3-6(7)8(11-12)9(13)14/h2-4H,1H3,(H,13,14)
Inchi Key OPJAPPYKVWTEQM-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-1-methylindazole-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-1-methylindazole-3-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart