APO945299013 Apollo Scientific Ltd

Tetrafluoropyrimidine

  • MDL number MFCD10687402
  • Cas Number : 767-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,4,5,6-Tetrafluoropyrimidine, 2,4,5,6-Tetrafluoro-1,3-diazine
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 83-85
Product Link https://store.apolloscientific.co.uk/product/tetrafluoropyrimidine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/tetrafluoropyrimidine
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart