APO945299023 Apollo Scientific Ltd

4-Amino-2,3-difluorobenzotrifluoride

  • MDL number MFCD00236206
  • Cas Number : 123950-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3-Difluoro-4-(trifluoromethyl)aniline
Purity 98
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-amino-23-difluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-23-difluorobenzotrifluoride
pictograms GHS07
hstatements H302;H312;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart