APO945299054 Apollo Scientific Ltd

3',5'-Dichloro-2,2,2,4'-tetrafluoroacetophenone

  • MDL number MFCD23382507
  • Cas Number : 1190865-44-1
  • Molecular Weight : 261

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(3,5-Dichloro-4-fluorophenyl)-2,2,2-trifluoroethan-1-one
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dichloro-2224-tetrafluoroacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloro-2224-tetrafluoroacetophenone
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart