APO945299062 Apollo Scientific Ltd

Potassium [3-(trifluoromethyl)benzyl]trifluoroborate

  • MDL number MFCD18712965
  • Cas Number : 1494466-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/potassium-3-trifluoromethylbenzyltrifluoroborate-95

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/potassium-3-trifluoromethylbenzyltrifluoroborate-95
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart