APO945299064 Apollo Scientific Ltd

Potassium (4-chloro-2-fluorobenzyl)trifluoroborate

  • MDL number MFCD30062988

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/potassium-4-chloro-2-fluorobenzyltrifluoroborate-95

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS https://store.apolloscientific.co.uk/product/potassium-4-chloro-2-fluorobenzyltrifluoroborate-95
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart