APO945299090 Apollo Scientific Ltd

4,5-Difluoro-2-methylaniline

  • MDL number MFCD08062378
  • Cas Number : 875664-57-6
  • Molecular Weight : 143.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-58
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-difluoro-2-methylaniline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/45-difluoro-2-methylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart